Learn More
Meso Scale Discovery U-PLEX Mouse IL-13 ASY
SDP
Catalog No. 5000078980
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-13 Assay Kit is a multiplex immunoassay for measuring mouse interleukin 13 (IL-13) in various biological samples It uses electrochemiluminescence (ECL) detection for high sensitivity and precision The kit includes a plate with capture antibodies ECL-tagged detection antibodies calibrators controls diluents buffers and seals It requires 50 L of sample per well and detects IL-13 from 0 05-1000 pg/mL with a total assay time of about 3 5 hours
Catalog No. 50-000-78980 Supplier Meso Scale Discovery Supplier No. K152UBK1
The Meso Scale Discovery U-PLEX Mouse IL-13 Assay Kit is a multiplex immunoassay for measuring mouse interleukin 13 (IL-13) in various biological samples It uses electrochemiluminescence (ECL) detection for high sensitivity and precision The kit includes a plate with capture antibodies ECL-tagged detection antibodies calibrators controls diluents buffers and seals It requires 50 L of sample per well and detects IL-13 from 0 05-1000 pg/mL with a total assay time of about 3 5 hours