Learn More
Meso Scale Discovery U-PLEX Mouse IL-13 ASY384-well
SDP
Catalog No. 5000081257
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-13 384-Well Assay is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 13 (IL-13) in cell culture supernatants serum plasma and other biological matrices This 10-plex assay allows simultaneous quantification of IL-13 and 9 other mouse cytokines and chemokines using minimal sample volume The assay utilizes MSD s patented SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with a broad dynamic range This product is suitable for mouse IL-13 detection in a high-throughput 384-well format
Catalog No. 50-000-81257 Supplier Meso Scale Discovery Supplier No. K252UBK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-13 384-Well Assay is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 13 (IL-13) in cell culture supernatants serum plasma and other biological matrices This 10-plex assay allows simultaneous quantification of IL-13 and 9 other mouse cytokines and chemokines using minimal sample volume The assay utilizes MSD s patented SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with a broad dynamic range This product is suitable for mouse IL-13 detection in a high-throughput 384-well format