Learn More
Meso Scale Discovery U-PLEX Mouse IL-13 ASYQP
SDP
Catalog No. 5000078699
Encompass
The MSD U-PLEX Mouse IL-13 Singleplex Assay is an electrochemiluminescence-based immunoassay kit designed for quantitative measurement of interleukin-13 in mouse biological samples This singleplex format allows focused detection of IL-13 a key cytokine involved in allergic responses and Th2 immune pathways The U-PLEX platform offers superior sensitivity wide dynamic range and minimal sample volume requirements compared to traditional ELISA methods The assay utilizes MSD s proprietary MULTI-SPOT technology for reliable reproducible results in research applications studying inflammation allergy and immune responses in mouse models
Catalog No. 50-000-78699 Supplier Meso Scale Discovery Supplier No. K152UBK22
The MSD U-PLEX Mouse IL-13 Singleplex Assay is an electrochemiluminescence-based immunoassay kit designed for quantitative measurement of interleukin-13 in mouse biological samples This singleplex format allows focused detection of IL-13 a key cytokine involved in allergic responses and Th2 immune pathways The U-PLEX platform offers superior sensitivity wide dynamic range and minimal sample volume requirements compared to traditional ELISA methods The assay utilizes MSD s proprietary MULTI-SPOT technology for reliable reproducible results in research applications studying inflammation allergy and immune responses in mouse models