Learn More
Meso Scale Discovery U-PLEX Mouse IL-15 ASYQP
SDP
Catalog No. 5000082157
Encompass
The MSD U-PLEX Mouse IL-15 Singleplex Assay is an electrochemiluminescence-based immunoassay kit designed for quantitative measurement of interleukin-15 protein levels in mouse biological samples This singleplex format allows focused analysis of IL-15 a cytokine involved in immune cell activation and proliferation The U-PLEX platform offers high sensitivity wide dynamic range and minimal sample volume requirements making it suitable for research applications in immunology inflammation studies and biomarker discovery
Catalog No. 50-000-82157 Supplier Meso Scale Discovery Supplier No. K152URK21
The MSD U-PLEX Mouse IL-15 Singleplex Assay is an electrochemiluminescence-based immunoassay kit designed for quantitative measurement of interleukin-15 protein levels in mouse biological samples This singleplex format allows focused analysis of IL-15 a cytokine involved in immune cell activation and proliferation The U-PLEX platform offers high sensitivity wide dynamic range and minimal sample volume requirements making it suitable for research applications in immunology inflammation studies and biomarker discovery