Learn More
Meso Scale Discovery U-PLEX Mouse IL-16 ASY 25 PL
SDP
Catalog No. 5000078357
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-16 Assay is a multiplex immunoassay kit that enables simultaneous quantification of mouse interleukin 16 (IL-16) in serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a wide dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators and controls Results are obtained in as little as 3 5 hours This assay allows for efficient reproducible analysis of IL-16 levels relevant to immunology and inflammation research
Catalog No. 50-000-78357 Supplier Meso Scale Discovery Supplier No. K152USK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-16 Assay is a multiplex immunoassay kit that enables simultaneous quantification of mouse interleukin 16 (IL-16) in serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a wide dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators and controls Results are obtained in as little as 3 5 hours This assay allows for efficient reproducible analysis of IL-16 levels relevant to immunology and inflammation research