Learn More
Meso Scale Discovery U-PLEX Mouse IL-17A ASY
SDP
Catalog No. 5000078368
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-17A Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 17A (IL-17A) in cell culture supernatants serum plasma and tissue lysates The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of mouse IL-17A with a lower limit of detection of 0 04 pg/mL and an upper limit of 10 000 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrators and sample diluent The assay takes approximately 3 5 hours to complete after a 2-hour incubation
Catalog No. 50-000-78368 Supplier Meso Scale Discovery Supplier No. K152UTK2
Add to Cart
Add to Cart
The Meso Scale Discovery U-PLEX Mouse IL-17A Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 17A (IL-17A) in cell culture supernatants serum plasma and tissue lysates The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of mouse IL-17A with a lower limit of detection of 0 04 pg/mL and an upper limit of 10 000 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrators and sample diluent The assay takes approximately 3 5 hours to complete after a 2-hour incubation