Learn More
Meso Scale Discovery U-PLEX Mouse IL-17A/F ASY
SDP
Catalog No. 5000078471
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-17A/F Assay is a multiplex immunoassay kit for quantifying mouse interleukin 17A and 17F in various biological matrices It uses electrochemiluminescence detection for high sensitivity and minimal cross-reactivity The assay requires 50 L or less sample volume and takes about 3 5 hours It includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent labels calibrators and assay diluent enabling efficient and accurate measurement of IL-17A and IL-17F in mouse samples
Catalog No. 50-000-78471 Supplier Meso Scale Discovery Supplier No. K152VYK2
Add to Cart
Add to Cart
The Meso Scale Discovery U-PLEX Mouse IL-17A/F Assay is a multiplex immunoassay kit for quantifying mouse interleukin 17A and 17F in various biological matrices It uses electrochemiluminescence detection for high sensitivity and minimal cross-reactivity The assay requires 50 L or less sample volume and takes about 3 5 hours It includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent labels calibrators and assay diluent enabling efficient and accurate measurement of IL-17A and IL-17F in mouse samples