Learn More
Meso Scale Discovery U-PLEX Mouse IL-17A/F ASYQP
SDP
Catalog No. 5000078713
Encompass
The U-PLEX Mouse IL-17A/F Assay (K152VYK) is a multiplex immunoassay kit from Meso Scale Discovery designed to simultaneously measure interleukin-17A and IL-17F cytokine levels in mouse biological samples This electrochemiluminescence-based assay provides high sensitivity detection with minimal sample volume requirements making it ideal for research applications studying inflammatory responses autoimmune diseases and immune system function in mouse models
Catalog No. 50-000-78713 Supplier Meso Scale Discovery Supplier No. K152VYK21
The U-PLEX Mouse IL-17A/F Assay (K152VYK) is a multiplex immunoassay kit from Meso Scale Discovery designed to simultaneously measure interleukin-17A and IL-17F cytokine levels in mouse biological samples This electrochemiluminescence-based assay provides high sensitivity detection with minimal sample volume requirements making it ideal for research applications studying inflammatory responses autoimmune diseases and immune system function in mouse models