Learn More
Meso Scale Discovery U-PLEX Mouse IL-17A/F ASYQP
SDP
Catalog No. 5000079008
Encompass
The U-PLEX Mouse IL-17A/F Assay (K152VYK) is a multiplex immunoassay kit designed for simultaneous quantitative measurement of interleukin-17A and IL-17F cytokines in mouse biological samples This electrochemiluminescence-based assay utilizes MSD s U-PLEX platform technology offering high sensitivity broad dynamic range and minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable cytokine analysis in research applications studying inflammatory responses and immune system function
Catalog No. 50-000-79008 Supplier Meso Scale Discovery Supplier No. K152VYK22
The U-PLEX Mouse IL-17A/F Assay (K152VYK) is a multiplex immunoassay kit designed for simultaneous quantitative measurement of interleukin-17A and IL-17F cytokines in mouse biological samples This electrochemiluminescence-based assay utilizes MSD s U-PLEX platform technology offering high sensitivity broad dynamic range and minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable cytokine analysis in research applications studying inflammatory responses and immune system function