Learn More
Meso Scale Discovery U-PLEX Mouse IL-17C ASY
SDP
Catalog No. 5000080333
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-17C Assay Kit is a multiplex immunoassay that enables simultaneous quantification of mouse interleukin 17C (IL-17C) in cell culture supernatants serum plasma and other biological matrices This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and quantify IL-17C concentrations This assay allows for rapid reproducible quantification of IL-17C from mouse samples
Catalog No. 50-000-80333 Supplier Meso Scale Discovery Supplier No. K152WJK2
The Meso Scale Discovery U-PLEX Mouse IL-17C Assay Kit is a multiplex immunoassay that enables simultaneous quantification of mouse interleukin 17C (IL-17C) in cell culture supernatants serum plasma and other biological matrices This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and quantify IL-17C concentrations This assay allows for rapid reproducible quantification of IL-17C from mouse samples