Learn More
Meso Scale Discovery U-PLEX Mouse IL-17C ASY
SDP
Catalog No. 5000082112
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-17C Assay is a multiplex immunoassay kit that enables simultaneous quantification of mouse interleukin 17C (IL-17C) in serum plasma and cell culture supernatants The assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay allows for the accurate and reliable measurement of IL-17C which plays an important role in mucosal immunity and inflammation With minimal hands-on time required this kit serves as a useful tool for mouse IL-17C detection in research studies involving immunology and inflammation
Catalog No. 50-000-82112 Supplier Meso Scale Discovery Supplier No. K152WJK1
The Meso Scale Discovery U-PLEX Mouse IL-17C Assay is a multiplex immunoassay kit that enables simultaneous quantification of mouse interleukin 17C (IL-17C) in serum plasma and cell culture supernatants The assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay allows for the accurate and reliable measurement of IL-17C which plays an important role in mucosal immunity and inflammation With minimal hands-on time required this kit serves as a useful tool for mouse IL-17C detection in research studies involving immunology and inflammation