Learn More
Meso Scale Discovery U-PLEX Mouse IL-17E/IL-25 ASY
SDP
Catalog No. 5000079009
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-17E/IL-25 Assay Kit is a multiplex immunoassay that measures mouse interleukin 17E (IL-17E) and interleukin 25 (IL-25) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This multiplex assay allows simultaneous quantification of IL-17E and IL-25 from a single sample in as little as 3 5 hours It requires just 25 L of sample and has a lower limit of detection of 0 04 pg/mL for IL-17E and 0 03 pg/mL for IL-25 The assay is for research use only
Catalog No. 50-000-79009 Supplier Meso Scale Discovery Supplier No. K152VZK2
The Meso Scale Discovery U-PLEX Mouse IL-17E/IL-25 Assay Kit is a multiplex immunoassay that measures mouse interleukin 17E (IL-17E) and interleukin 25 (IL-25) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This multiplex assay allows simultaneous quantification of IL-17E and IL-25 from a single sample in as little as 3 5 hours It requires just 25 L of sample and has a lower limit of detection of 0 04 pg/mL for IL-17E and 0 03 pg/mL for IL-25 The assay is for research use only