Learn More
Meso Scale Discovery U-PLEX Mouse IL-17E/IL-25 ASY
SDP
Catalog No. 5000079365
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-17E/IL-25 Assay Kit quantifies mouse interleukin 17E and 25 in serum plasma and cell culture supernatants using electrochemiluminescent detection It employs MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results from small samples This multiplex assay detects IL-17E and IL-25 simultaneously in one well aiding immune response monitoring The kit includes a 96-well plate with capture antibodies detection antibodies buffers blocker solutions SULFO-TAG labeled detection antibodies calibrators and controls
Catalog No. 50-000-79365 Supplier Meso Scale Discovery Supplier No. K152VZK1
The Meso Scale Discovery U-PLEX Mouse IL-17E/IL-25 Assay Kit quantifies mouse interleukin 17E and 25 in serum plasma and cell culture supernatants using electrochemiluminescent detection It employs MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results from small samples This multiplex assay detects IL-17E and IL-25 simultaneously in one well aiding immune response monitoring The kit includes a 96-well plate with capture antibodies detection antibodies buffers blocker solutions SULFO-TAG labeled detection antibodies calibrators and controls