Learn More
Meso Scale Discovery U-PLEX Mouse IL-1B ASY
SDP
Catalog No. 5000078349
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-1beta Assay is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 1 beta (IL-1beta) in cell culture supernatants serum plasma and other biological samples This assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The lower limit of detection is 0 04 pg/mL This assay is for research use only and not for use in diagnostic procedures It is available as a kit with plates and reagents for 96 tests
Catalog No. 50-000-78349 Supplier Meso Scale Discovery Supplier No. K152TUK2
The Meso Scale Discovery U-PLEX Mouse IL-1beta Assay is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 1 beta (IL-1beta) in cell culture supernatants serum plasma and other biological samples This assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The lower limit of detection is 0 04 pg/mL This assay is for research use only and not for use in diagnostic procedures It is available as a kit with plates and reagents for 96 tests