Learn More
Meso Scale Discovery U-PLEX Mouse IL-1B ASY384-well
SDP
Catalog No. 5000081216
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-1beta 384-Well Assay Kit is a multiplex immunoassay that measures mouse interleukin-1 beta (IL-1beta) in serum plasma cell culture supernatant and other biological fluids It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption in a 384-well plate format This assay kit includes a mouse IL-1beta standard and all necessary reagents and plates to perform the immunoassay It can be used for mouse cytokine profiling and inflammation studies With a testing time of just 3 5 hours this kit enables efficient and high-throughput quantification of IL-1beta
Catalog No. 50-000-81216 Supplier Meso Scale Discovery Supplier No. K252TUK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-1beta 384-Well Assay Kit is a multiplex immunoassay that measures mouse interleukin-1 beta (IL-1beta) in serum plasma cell culture supernatant and other biological fluids It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption in a 384-well plate format This assay kit includes a mouse IL-1beta standard and all necessary reagents and plates to perform the immunoassay It can be used for mouse cytokine profiling and inflammation studies With a testing time of just 3 5 hours this kit enables efficient and high-throughput quantification of IL-1beta