Learn More
Meso Scale Discovery U-PLEX Mouse IL-2 ASY 25 PL
SDP
Catalog No. 5000078351
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-2 Assay Kit is a sensitive immunoassay for detecting and quantifying mouse interleukin 2 (IL-2) in various biological fluids It uses MSD s electrochemiluminescent detection technology and includes plates with capture antibodies detection antibodies and calibrators The assay has a sensitivity of 0 06 pg/mL a dynamic range over 3 logs requires 50 L sample volume and is compatible with MSD platforms The kit contains all necessary reagents for measuring mouse IL-2
Catalog No. 50-000-78351 Supplier Meso Scale Discovery Supplier No. K152TVK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-2 Assay Kit is a sensitive immunoassay for detecting and quantifying mouse interleukin 2 (IL-2) in various biological fluids It uses MSD s electrochemiluminescent detection technology and includes plates with capture antibodies detection antibodies and calibrators The assay has a sensitivity of 0 06 pg/mL a dynamic range over 3 logs requires 50 L sample volume and is compatible with MSD platforms The kit contains all necessary reagents for measuring mouse IL-2