Learn More
Meso Scale Discovery U-PLEX Mouse IL-2 ASY
SDP
Catalog No. 5000078436
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-2 Assay Kit is a sensitive immunoassay for detecting and quantifying mouse interleukin 2 (IL-2) in various biological fluids It uses MSD s electrochemiluminescent technology and includes plates with capture antibodies detection antibodies calibrators and buffers for 96 tests With a sensitivity of 0 06 pg/mL and a range of 0 06-1500 pg/mL it requires only 25 L of sample and provides results in 3 5 hours Ideal for mouse vaccine studies immunotherapy research and cytokine profiling
Catalog No. 50-000-78436 Supplier Meso Scale Discovery Supplier No. K152TVK1
The Meso Scale Discovery U-PLEX Mouse IL-2 Assay Kit is a sensitive immunoassay for detecting and quantifying mouse interleukin 2 (IL-2) in various biological fluids It uses MSD s electrochemiluminescent technology and includes plates with capture antibodies detection antibodies calibrators and buffers for 96 tests With a sensitivity of 0 06 pg/mL and a range of 0 06-1500 pg/mL it requires only 25 L of sample and provides results in 3 5 hours Ideal for mouse vaccine studies immunotherapy research and cytokine profiling