Learn More
Meso Scale Discovery U-PLEX Mouse IL-2 ASY384-well
SDP
Catalog No. 5000078973
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-2 384-Well Assay Kit is a multiplex immunoassay for measuring mouse interleukin 2 (IL-2) in various biological fluids It uses electrochemiluminescence (ECL) detection offering high sensitivity a 5-log dynamic range and low sample use in a 384-well format Requiring 10 L per well it completes in 3 5 hours The kit includes a plate with capture antibodies ECL-tagged detection antibodies and calibrators for a standard curve It tests 40 samples in duplicate with reagents for one plate
Catalog No. 50-000-78973 Supplier Meso Scale Discovery Supplier No. K252TVK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-2 384-Well Assay Kit is a multiplex immunoassay for measuring mouse interleukin 2 (IL-2) in various biological fluids It uses electrochemiluminescence (ECL) detection offering high sensitivity a 5-log dynamic range and low sample use in a 384-well format Requiring 10 L per well it completes in 3 5 hours The kit includes a plate with capture antibodies ECL-tagged detection antibodies and calibrators for a standard curve It tests 40 samples in duplicate with reagents for one plate