Learn More
Meso Scale Discovery U-PLEX Mouse IL-21 ASY 25 PL
SDP
Catalog No. 5000080419
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-21 Assay Kit is a multiplex immunoassay that allows simultaneous measurement of mouse interleukin 21 (IL-21) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrator diluent wash buffer read buffer and a vial of lyophilized calibrator containing recombinant mouse IL-21 The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL for mouse IL-21
Catalog No. 50-000-80419 Supplier Meso Scale Discovery Supplier No. K152WBK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-21 Assay Kit is a multiplex immunoassay that allows simultaneous measurement of mouse interleukin 21 (IL-21) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrator diluent wash buffer read buffer and a vial of lyophilized calibrator containing recombinant mouse IL-21 The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL for mouse IL-21