Learn More
Meso Scale Discovery U-PLEX Mouse IL-21 ASY384-well
SDP
Catalog No. 5000081314
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-21 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 21 (IL-21) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of mouse IL-21 with a lower limit of detection of 1 15 pg/mL and an upper limit of quantification of 2500 pg/mL The kit includes a mouse IL-21 calibrator and pre-coated plates to enable high-throughput analysis of mouse IL-21 levels
Catalog No. 50-000-81314 Supplier Meso Scale Discovery Supplier No. K252WBK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-21 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 21 (IL-21) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of mouse IL-21 with a lower limit of detection of 1 15 pg/mL and an upper limit of quantification of 2500 pg/mL The kit includes a mouse IL-21 calibrator and pre-coated plates to enable high-throughput analysis of mouse IL-21 levels