Learn More
Meso Scale Discovery U-PLEX Mouse IL-22 ASY 25 PL
SDP
Catalog No. 5000078689
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-22 Assay Kit is a multiplex immunoassay for measuring mouse interleukin 22 (IL-22) in various samples It uses electrochemiluminescence detection for high sensitivity and precision The kit includes a plate with capture antibodies ECL-tagged detection antibodies calibrators controls and necessary buffers It requires 50 L of sample per well and quantifies IL-22 from 1 5 to 10 000 pg/mL in about 3 5 hours aiding in the study of mucosal immunity tissue regeneration and inflammation
Catalog No. 50-000-78689 Supplier Meso Scale Discovery Supplier No. K152WIK4
Add to Cart
Add to Cart
Request a Quote
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Meso Scale Discovery U-PLEX Mouse IL-22 Assay Kit is a multiplex immunoassay for measuring mouse interleukin 22 (IL-22) in various samples It uses electrochemiluminescence detection for high sensitivity and precision The kit includes a plate with capture antibodies ECL-tagged detection antibodies calibrators controls and necessary buffers It requires 50 L of sample per well and quantifies IL-22 from 1 5 to 10 000 pg/mL in about 3 5 hours aiding in the study of mucosal immunity tissue regeneration and inflammation