Learn More
Meso Scale Discovery U-PLEX Mouse IL-22 ASY
SDP
Catalog No. 5000078686
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-22 Assay Kit is a multiplex immunoassay for measuring mouse interleukin 22 (IL-22) in various biological samples It uses electrochemiluminescence detection for high sensitivity and precision The kit includes necessary reagents and requires 50 L of sample per well quantifying IL-22 between 1 5 and 10 000 pg/mL in about 3 5 hours This assay aids in studying IL-22 s roles in mucosal immunity tissue regeneration and inflammation
Catalog No. 50-000-78686 Supplier Meso Scale Discovery Supplier No. K152WIK1
The Meso Scale Discovery U-PLEX Mouse IL-22 Assay Kit is a multiplex immunoassay for measuring mouse interleukin 22 (IL-22) in various biological samples It uses electrochemiluminescence detection for high sensitivity and precision The kit includes necessary reagents and requires 50 L of sample per well quantifying IL-22 between 1 5 and 10 000 pg/mL in about 3 5 hours This assay aids in studying IL-22 s roles in mucosal immunity tissue regeneration and inflammation