Learn More
Meso Scale Discovery U-PLEX Mouse IL-22 ASY384-well
SDP
Catalog No. 5000078955
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-22 384-Well Assay Kit is a multiplex immunoassay that measures mouse interleukin 22 (IL-22) in serum plasma cell culture supernatants and other biological fluids It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra-/inter-assay precision This 384-well plate assay allows high throughput screening and only requires 10-25 L sample volume per well The kit includes a plate coated with capture antibodies detection antibodies read buffer wash buffer diluent and calibrators to generate a standard curve
Catalog No. 50-000-78955 Supplier Meso Scale Discovery Supplier No. K252WIK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-22 384-Well Assay Kit is a multiplex immunoassay that measures mouse interleukin 22 (IL-22) in serum plasma cell culture supernatants and other biological fluids It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra-/inter-assay precision This 384-well plate assay allows high throughput screening and only requires 10-25 L sample volume per well The kit includes a plate coated with capture antibodies detection antibodies read buffer wash buffer diluent and calibrators to generate a standard curve