Learn More
Meso Scale Discovery U-PLEX Mouse IL-22 ASYQP
SDP
Catalog No. 5000078687
Encompass
The U-PLEX Mouse IL-22 Assay (K152WIK) is a highly sensitive immunoassay kit designed to quantitatively measure interleukin-22 levels in mouse biological samples This electrochemiluminescence-based assay offers superior sensitivity and precision for cytokine detection in research applications The kit provides reliable reproducible results for studying IL-22 s role in immune responses inflammation and tissue homeostasis in mouse models making it essential for immunology and inflammatory disease research
Catalog No. 50-000-78687 Supplier Meso Scale Discovery Supplier No. K152WIK22
The U-PLEX Mouse IL-22 Assay (K152WIK) is a highly sensitive immunoassay kit designed to quantitatively measure interleukin-22 levels in mouse biological samples This electrochemiluminescence-based assay offers superior sensitivity and precision for cytokine detection in research applications The kit provides reliable reproducible results for studying IL-22 s role in immune responses inflammation and tissue homeostasis in mouse models making it essential for immunology and inflammatory disease research