Learn More
Meso Scale Discovery U-PLEX Mouse IL-23 ASY384-well
SDP
Catalog No. 5000081204
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-23 384-Well Assay Kit is a multiplex immunoassay for quantifying mouse interleukin 23 (IL-23) in various biological matrices It uses MSD s SULFO-TAG labels offering high sensitivity broad dynamic range and low sample volume The kit includes a 384-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are analyzed on MSD s platform providing pg/mL IL-23 concentrations enabling high-throughput measurement from mouse samples
Catalog No. 50-000-81204 Supplier Meso Scale Discovery Supplier No. K252WGK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-23 384-Well Assay Kit is a multiplex immunoassay for quantifying mouse interleukin 23 (IL-23) in various biological matrices It uses MSD s SULFO-TAG labels offering high sensitivity broad dynamic range and low sample volume The kit includes a 384-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are analyzed on MSD s platform providing pg/mL IL-23 concentrations enabling high-throughput measurement from mouse samples