Learn More
Meso Scale Discovery U-PLEX Mouse IL-31 ASY 25 PL
SDP
Catalog No. 5000079056
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-31 Assay Kit is a multiplex immunoassay that measures mouse interleukin 31 (IL-31) in serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrator diluent wash buffer read buffer and a vial of lyophilized calibrator that must be reconstituted before use The assay can quantify mouse IL-31 concentrations between 1 5 and 10 000 pg/mL
Catalog No. 50-000-79056 Supplier Meso Scale Discovery Supplier No. K152WEK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-31 Assay Kit is a multiplex immunoassay that measures mouse interleukin 31 (IL-31) in serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrator diluent wash buffer read buffer and a vial of lyophilized calibrator that must be reconstituted before use The assay can quantify mouse IL-31 concentrations between 1 5 and 10 000 pg/mL