Learn More
Meso Scale Discovery U-PLEX Mouse IL-31 ASY
SDP
Catalog No. 5000082111
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-31 Assay Kit quantifies mouse interleukin 31 (IL-31) in serum plasma and other samples This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection of IL-31 at sub-picogram levels It includes a 96-well plate with capture antibody detection antibody with SULFO-TAG calibrators and diluent The assay runs on MSD s SECTOR Imager providing results in 3 5 hours ensuring reliable quantification of mouse IL-31
Catalog No. 50-000-82111 Supplier Meso Scale Discovery Supplier No. K152WEK2
The Meso Scale Discovery U-PLEX Mouse IL-31 Assay Kit quantifies mouse interleukin 31 (IL-31) in serum plasma and other samples This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection of IL-31 at sub-picogram levels It includes a 96-well plate with capture antibody detection antibody with SULFO-TAG calibrators and diluent The assay runs on MSD s SECTOR Imager providing results in 3 5 hours ensuring reliable quantification of mouse IL-31