Learn More
Meso Scale Discovery U-PLEX Mouse IL-31 ASY384-well
SDP
Catalog No. 5000080181
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-31 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 31 (IL-31) in serum plasma and cell culture supernatants The assay uses MSD GOLD Streptavidin Multi-Array plates and a SULFO-TAG labeled detection antibody The kit includes a plate detection antibody calibrator and assay diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL The total assay time is 3 5 hours This product can be used to measure mouse IL-31 levels in immune response studies
Catalog No. 50-000-80181 Supplier Meso Scale Discovery Supplier No. K252WEK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-31 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 31 (IL-31) in serum plasma and cell culture supernatants The assay uses MSD GOLD Streptavidin Multi-Array plates and a SULFO-TAG labeled detection antibody The kit includes a plate detection antibody calibrator and assay diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL The total assay time is 3 5 hours This product can be used to measure mouse IL-31 levels in immune response studies