Learn More
Meso Scale Discovery U-PLEX Mouse IL-33 ASY
SDP
Catalog No. 5000082243
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-33 Assay Kit is a multiplex immunoassay that measures mouse interleukin 33 (IL-33) in serum plasma cell culture supernatants and other biological fluids This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and an MSD read buffer Sample incubation time is just 2 hours The kit can test up to 80 samples in duplicate and has a lower limit of detection of 0 04 pg/mL for mouse IL-33
Catalog No. 50-000-82243 Supplier Meso Scale Discovery Supplier No. K152WFK1
The Meso Scale Discovery U-PLEX Mouse IL-33 Assay Kit is a multiplex immunoassay that measures mouse interleukin 33 (IL-33) in serum plasma cell culture supernatants and other biological fluids This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and an MSD read buffer Sample incubation time is just 2 hours The kit can test up to 80 samples in duplicate and has a lower limit of detection of 0 04 pg/mL for mouse IL-33