Learn More
Meso Scale Discovery U-PLEX Mouse IL-33 ASY
SDP
Catalog No. 5000082409
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-33 Assay Kit is a multiplex immunoassay for detecting and quantifying mouse interleukin 33 (IL-33) in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies calibrator diluent wash buffer read buffer and a 10-point standard curve The assay takes about 3 5 hours and is compatible with lab automation systems enhancing biomarker research efficiency
Catalog No. 50-000-82409 Supplier Meso Scale Discovery Supplier No. K152WFK2
The Meso Scale Discovery U-PLEX Mouse IL-33 Assay Kit is a multiplex immunoassay for detecting and quantifying mouse interleukin 33 (IL-33) in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies calibrator diluent wash buffer read buffer and a 10-point standard curve The assay takes about 3 5 hours and is compatible with lab automation systems enhancing biomarker research efficiency