Learn More
Meso Scale Discovery U-PLEX Mouse IL-33 ASY384-well
SDP
Catalog No. 5000081188
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-33 384-Well Assay Kit is a 384-well multiplex immunoassay that measures mouse interleukin-33 (IL-33) in serum plasma cell culture supernatants cell and tissue lysates and other biological samples The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with minimal matrix interference This multiplex kit includes a plate coated with capture antibodies detection antibodies calibrators and controls needed to quantify IL-33 levels accurately The assay requires a small sample volume and provides results in as little as 3 5 hours
Catalog No. 50-000-81188 Supplier Meso Scale Discovery Supplier No. K252WFK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-33 384-Well Assay Kit is a 384-well multiplex immunoassay that measures mouse interleukin-33 (IL-33) in serum plasma cell culture supernatants cell and tissue lysates and other biological samples The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with minimal matrix interference This multiplex kit includes a plate coated with capture antibodies detection antibodies calibrators and controls needed to quantify IL-33 levels accurately The assay requires a small sample volume and provides results in as little as 3 5 hours