Learn More
Meso Scale Discovery U-PLEX Mouse IL-33 ASYQP
SDP
Catalog No. 5000079057
Encompass
The U-PLEX Mouse IL-33 384-Well Assay (K252WFK) is a highly sensitive immunoassay designed to quantify interleukin-33 levels in mouse samples This multiplex platform utilizes electrochemiluminescence detection technology to deliver superior sensitivity and dynamic range compared to traditional ELISA methods The 384-well format enables high-throughput screening capabilities making it ideal for large-scale research studies The assay provides reliable reproducible results for measuring IL-33 a key cytokine involved in immune responses and inflammatory processes in mouse models
Catalog No. 50-000-79057 Supplier Meso Scale Discovery Supplier No. K152WFK21
The U-PLEX Mouse IL-33 384-Well Assay (K252WFK) is a highly sensitive immunoassay designed to quantify interleukin-33 levels in mouse samples This multiplex platform utilizes electrochemiluminescence detection technology to deliver superior sensitivity and dynamic range compared to traditional ELISA methods The 384-well format enables high-throughput screening capabilities making it ideal for large-scale research studies The assay provides reliable reproducible results for measuring IL-33 a key cytokine involved in immune responses and inflammatory processes in mouse models