Learn More
Meso Scale Discovery U-PLEX Mouse IL-4 ASY
SDP
Catalog No. 5000078982
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-4 Assay Kit is a sensitive immunoassay for detecting and quantifying mouse interleukin 4 (IL-4) in samples like serum plasma and cell culture supernatant It uses MSD s electrochemiluminescent technology offering high sensitivity a wide dynamic range and quick results The assay detects IL-4 at 0 03 pg/mL with a 7-log range The kit includes a 96-well plate with capture antibodies detection antibodies and calibrators The assay completes in 3 5 hours after a 2-hour incubation providing a robust tool for research
Catalog No. 50-000-78982 Supplier Meso Scale Discovery Supplier No. K152TWK1
The Meso Scale Discovery U-PLEX Mouse IL-4 Assay Kit is a sensitive immunoassay for detecting and quantifying mouse interleukin 4 (IL-4) in samples like serum plasma and cell culture supernatant It uses MSD s electrochemiluminescent technology offering high sensitivity a wide dynamic range and quick results The assay detects IL-4 at 0 03 pg/mL with a 7-log range The kit includes a 96-well plate with capture antibodies detection antibodies and calibrators The assay completes in 3 5 hours after a 2-hour incubation providing a robust tool for research