Learn More
Meso Scale Discovery U-PLEX Mouse IL-4 ASY 25 PL
SDP
Catalog No. 5000078306
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-4 Assay Kit is an electrochemiluminescent immunoassay for quantifying mouse interleukin 4 (IL-4) in various samples It uses MSD s SECTOR Imager to detect signals from microplate wells The kit includes a capture antibody for IL-4 and a ruthenium-labeled detection antibody It has a detection range of 0 05-1000 pg/mL with no cross-reactivity with other cytokines The kit provides a 96-well plate detection antibody calibrators 10X diluent and components for 96 tests
Catalog No. 50-000-78306 Supplier Meso Scale Discovery Supplier No. K152TWK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-4 Assay Kit is an electrochemiluminescent immunoassay for quantifying mouse interleukin 4 (IL-4) in various samples It uses MSD s SECTOR Imager to detect signals from microplate wells The kit includes a capture antibody for IL-4 and a ruthenium-labeled detection antibody It has a detection range of 0 05-1000 pg/mL with no cross-reactivity with other cytokines The kit provides a 96-well plate detection antibody calibrators 10X diluent and components for 96 tests