Learn More
Meso Scale Discovery U-PLEX Mouse IL-4 ASY
SDP
Catalog No. 5000079360
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-4 Assay Kit is a sensitive immunoassay for detecting and quantifying mouse interleukin 4 (IL-4) in samples like serum plasma and cell culture supernatant It uses MSD s electrochemiluminescent technology offering high sensitivity a wide dynamic range and quick results The assay detects IL-4 at 0 03 pg/mL with a 7-log range The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and necessary reagents Sample incubation is 2 hours enabling simple and reliable IL-4 measurement for research
Catalog No. 50-000-79360 Supplier Meso Scale Discovery Supplier No. K152TWK2
The Meso Scale Discovery U-PLEX Mouse IL-4 Assay Kit is a sensitive immunoassay for detecting and quantifying mouse interleukin 4 (IL-4) in samples like serum plasma and cell culture supernatant It uses MSD s electrochemiluminescent technology offering high sensitivity a wide dynamic range and quick results The assay detects IL-4 at 0 03 pg/mL with a 7-log range The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and necessary reagents Sample incubation is 2 hours enabling simple and reliable IL-4 measurement for research