Learn More
Meso Scale Discovery U-PLEX Mouse IL-4 ASY384-well
SDP
Catalog No. 5000081529
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-4 384-Well Assay Kit is a multiplex immunoassay for measuring mouse interleukin 4 (IL-4) in various biological samples It uses electrochemiluminescence detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 384-well plate with capture and detection antibodies calibrators and controls It runs in about 3 5 hours testing up to 384 samples in duplicate with data analysis via MSD Discovery Workbench software for accurate IL-4 quantification
Catalog No. 50-000-81529 Supplier Meso Scale Discovery Supplier No. K252TWK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-4 384-Well Assay Kit is a multiplex immunoassay for measuring mouse interleukin 4 (IL-4) in various biological samples It uses electrochemiluminescence detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 384-well plate with capture and detection antibodies calibrators and controls It runs in about 3 5 hours testing up to 384 samples in duplicate with data analysis via MSD Discovery Workbench software for accurate IL-4 quantification