Learn More
Meso Scale Discovery U-PLEX Mouse IL-5 ASY 25 PL
SDP
Catalog No. 5000079366
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-5 Assay Kit is a multiplex immunoassay for measuring mouse interleukin 5 (IL-5) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity and precision The kit includes a plate with capture antibodies ECL-tagged detection antibodies calibrators and diluent The assay takes about 3 5 hours and requires a Meso Scale Discovery instrument like the MESO QuickPlex SQ 120 It allows accurate quantification of mouse IL-5 with minimal sample volume and a broad dynamic range
Catalog No. 50-000-79366 Supplier Meso Scale Discovery Supplier No. K152UOK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-5 Assay Kit is a multiplex immunoassay for measuring mouse interleukin 5 (IL-5) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity and precision The kit includes a plate with capture antibodies ECL-tagged detection antibodies calibrators and diluent The assay takes about 3 5 hours and requires a Meso Scale Discovery instrument like the MESO QuickPlex SQ 120 It allows accurate quantification of mouse IL-5 with minimal sample volume and a broad dynamic range