Learn More
Meso Scale Discovery U-PLEX Mouse IL-5 ASY
SDP
Catalog No. 5000078468
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-5 Assay Kit is an electrochemiluminescent immunoassay for quantifying mouse interleukin 5 (IL-5) in serum plasma and cell culture supernatants It uses MSD s SECTOR Imager detecting IL-5 from 0 04 to 1000 pg/mL The kit includes a 96-well plate with capture antibody detection antibody with MSD SULFO-TAG calibrators and diluent The assay takes 3 5 hours and supports research on mouse models of asthma allergies and other IL-5-related immunological diseases
Catalog No. 50-000-78468 Supplier Meso Scale Discovery Supplier No. K152UOK1
The Meso Scale Discovery U-PLEX Mouse IL-5 Assay Kit is an electrochemiluminescent immunoassay for quantifying mouse interleukin 5 (IL-5) in serum plasma and cell culture supernatants It uses MSD s SECTOR Imager detecting IL-5 from 0 04 to 1000 pg/mL The kit includes a 96-well plate with capture antibody detection antibody with MSD SULFO-TAG calibrators and diluent The assay takes 3 5 hours and supports research on mouse models of asthma allergies and other IL-5-related immunological diseases