Learn More
Meso Scale Discovery U-PLEX Mouse IL-5 ASYQP
SDP
Catalog No. 5000078715
Encompass
MSD s U-PLEX mouse IL-5 assay is an electrochemiluminescence-based immunoassay for quantitative measurement of interleukin-5 in mouse samples This multiplex platform allows researchers to measure IL-5 alongside other analytes in a single well providing efficient and sensitive detection for cytokine research applications The U-PLEX technology offers flexibility for custom panel creation and requires minimal sample volume while delivering reliable reproducible results for mouse studies
Catalog No. 50-000-78715 Supplier Meso Scale Discovery Supplier No. K152UOK22
MSD s U-PLEX mouse IL-5 assay is an electrochemiluminescence-based immunoassay for quantitative measurement of interleukin-5 in mouse samples This multiplex platform allows researchers to measure IL-5 alongside other analytes in a single well providing efficient and sensitive detection for cytokine research applications The U-PLEX technology offers flexibility for custom panel creation and requires minimal sample volume while delivering reliable reproducible results for mouse studies