Learn More
Meso Scale Discovery U-PLEX Mouse IL-5 ASYQP
SDP
Catalog No. 5000080441
Encompass
The MSD U-PLEX Mouse IL-5 Singleplex Assay is an electrochemiluminescence-based immunoassay kit designed for quantitative measurement of interleukin-5 in mouse biological samples This singleplex format allows focused detection of IL-5 a key cytokine involved in eosinophil activation and allergic responses The U-PLEX platform offers superior sensitivity wide dynamic range and minimal sample volume requirements compared to traditional ELISA methods
Catalog No. 50-000-80441 Supplier Meso Scale Discovery Supplier No. K152UOK21
The MSD U-PLEX Mouse IL-5 Singleplex Assay is an electrochemiluminescence-based immunoassay kit designed for quantitative measurement of interleukin-5 in mouse biological samples This singleplex format allows focused detection of IL-5 a key cytokine involved in eosinophil activation and allergic responses The U-PLEX platform offers superior sensitivity wide dynamic range and minimal sample volume requirements compared to traditional ELISA methods