Learn More
Meso Scale Discovery U-PLEX Mouse IL-6 ASY 25 PL
SDP
Catalog No. 5000078352
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-6 Assay Kit is a sensitive immunoassay for detecting mouse interleukin 6 (IL-6) in various samples It uses MSD s electrochemiluminescent technology offering high sensitivity a wide dynamic range and quick results The kit includes a 96-well plate with capture antibodies for mouse IL-6 detection antibodies with an electrochemiluminescent label and calibrators for a standard curve Sample incubation is 2 hours with no wash steps quantifying IL-6 from 0 2-10 000 pg/mL
Catalog No. 50-000-78352 Supplier Meso Scale Discovery Supplier No. K152TXK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-6 Assay Kit is a sensitive immunoassay for detecting mouse interleukin 6 (IL-6) in various samples It uses MSD s electrochemiluminescent technology offering high sensitivity a wide dynamic range and quick results The kit includes a 96-well plate with capture antibodies for mouse IL-6 detection antibodies with an electrochemiluminescent label and calibrators for a standard curve Sample incubation is 2 hours with no wash steps quantifying IL-6 from 0 2-10 000 pg/mL