Learn More
Meso Scale Discovery U-PLEX Mouse IL-6 ASY
SDP
Catalog No. 5000078643
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-6 Assay Kit is a multiplex immunoassay for measuring mouse interleukin 6 (IL-6) in various biological samples It uses electrochemiluminescence (ECL) detection for high sensitivity and precision The kit includes a 96-well plate with capture antibodies ECL-tagged detection antibodies calibrators and diluent The assay takes about 3 5 hours and requires a Meso Scale Discovery instrument It detects IL-6 with a lower limit of 0 3 pg/mL allowing quantitation across a wide range
Catalog No. 50-000-78643 Supplier Meso Scale Discovery Supplier No. K152TXK1
The Meso Scale Discovery U-PLEX Mouse IL-6 Assay Kit is a multiplex immunoassay for measuring mouse interleukin 6 (IL-6) in various biological samples It uses electrochemiluminescence (ECL) detection for high sensitivity and precision The kit includes a 96-well plate with capture antibodies ECL-tagged detection antibodies calibrators and diluent The assay takes about 3 5 hours and requires a Meso Scale Discovery instrument It detects IL-6 with a lower limit of 0 3 pg/mL allowing quantitation across a wide range