Learn More
Meso Scale Discovery U-PLEX Mouse IL-6 ASY384-well
SDP
Catalog No. 5000078956
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-6 384-Well Assay Kit is a multiplex immunoassay that measures mouse interleukin 6 (IL-6) in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra-/inter-assay precision The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator solutions It allows simultaneous measurement of mouse IL-6 along with other biomarkers in the same sample The assay takes approximately 3 hours to complete
Catalog No. 50-000-78956 Supplier Meso Scale Discovery Supplier No. K252TXK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-6 384-Well Assay Kit is a multiplex immunoassay that measures mouse interleukin 6 (IL-6) in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra-/inter-assay precision The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator solutions It allows simultaneous measurement of mouse IL-6 along with other biomarkers in the same sample The assay takes approximately 3 hours to complete