Learn More
Meso Scale Discovery U-PLEX Mouse IL-9 ASY 25 PL
SDP
Catalog No. 5000079108
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-9 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 9 (IL-9) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of mouse IL-9 with a lower limit of detection of 0 3 pg/mL and an upper limit of 10 000 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrators and sample diluent The assay takes approximately 3 5 hours to complete after a 2 hour incubation
Catalog No. 50-000-79108 Supplier Meso Scale Discovery Supplier No. K152XKK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-9 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 9 (IL-9) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of mouse IL-9 with a lower limit of detection of 0 3 pg/mL and an upper limit of 10 000 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrators and sample diluent The assay takes approximately 3 5 hours to complete after a 2 hour incubation