Learn More
Meso Scale Discovery U-PLEX Mouse IL-9 ASY
SDP
Catalog No. 5000079106
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-9 Assay Kit is a multiplex immunoassay that measures mouse interleukin 9 (IL-9) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and diluents It can accurately quantify IL-9 concentrations from as low as 24 pg/mL to 50 000 pg/mL The assay takes 3 5 hours to complete including a 2 hour incubation step
Catalog No. 50-000-79106 Supplier Meso Scale Discovery Supplier No. K152XKK1
The Meso Scale Discovery U-PLEX Mouse IL-9 Assay Kit is a multiplex immunoassay that measures mouse interleukin 9 (IL-9) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and diluents It can accurately quantify IL-9 concentrations from as low as 24 pg/mL to 50 000 pg/mL The assay takes 3 5 hours to complete including a 2 hour incubation step