Learn More
Meso Scale Discovery U-PLEX Mouse IL-9 ASY384-well
SDP
Catalog No. 5000078953
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-9 384-Well Assay Kit is a multiplex immunoassay that measures mouse interleukin 9 (IL-9) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and buffers for 384 tests It can accurately quantify IL-9 concentrations from 50 - 500 000 pg/mL with intra- and inter-assay precision of 10% CV The assay takes 3 5 hours to complete
Catalog No. 50-000-78953 Supplier Meso Scale Discovery Supplier No. K252XKK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-9 384-Well Assay Kit is a multiplex immunoassay that measures mouse interleukin 9 (IL-9) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and buffers for 384 tests It can accurately quantify IL-9 concentrations from 50 - 500 000 pg/mL with intra- and inter-assay precision of 10% CV The assay takes 3 5 hours to complete