Learn More
Meso Scale Discovery U-PLEX Mouse Insulin ASY
SDP
Catalog No. 5000081447
Encompass
The Meso Scale Discovery U-PLEX Mouse Insulin Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of insulin in mouse serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to perform the assay The kit has a broad dynamic range of 0 05-1000 pg/mL and exhibits no cross-reactivity with mouse C-peptide The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-81447 Supplier Meso Scale Discovery Supplier No. K1526HK1
The Meso Scale Discovery U-PLEX Mouse Insulin Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of insulin in mouse serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to perform the assay The kit has a broad dynamic range of 0 05-1000 pg/mL and exhibits no cross-reactivity with mouse C-peptide The assay takes approximately 3 5 hours to complete