Learn More
Meso Scale Discovery U-PLEX Mouse Insulin ASY
SDP
Catalog No. 5000082383
Encompass
The Meso Scale Discovery U-PLEX Mouse Insulin Assay Kit is an electrochemiluminescent immunoassay for measuring insulin in mouse serum plasma and cell culture supernatants It uses SULFO-TAG labels and multi-array technology for sensitive reproducible results with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are analyzed on the QuickPlex SQ 120 instrument with high specificity for mouse insulin and minimal cross-reactivity
Catalog No. 50-000-82383 Supplier Meso Scale Discovery Supplier No. K1526HK2
The Meso Scale Discovery U-PLEX Mouse Insulin Assay Kit is an electrochemiluminescent immunoassay for measuring insulin in mouse serum plasma and cell culture supernatants It uses SULFO-TAG labels and multi-array technology for sensitive reproducible results with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are analyzed on the QuickPlex SQ 120 instrument with high specificity for mouse insulin and minimal cross-reactivity