Learn More
Meso Scale Discovery U-PLEX Mouse Insulin ASYQP
SDP
Catalog No. 5000081379
Encompass
The Meso Scale Discovery U-PLEX Mouse Insulin Assay is an electrochemiluminescent immunoassay for measuring insulin in mouse serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive accurate results in 3 5 hours The assay detects as low as 50 pg/mL and includes a calibrator for a standard curve from 2-10 000 pg/mL It requires 25 L of sample and offers high specificity without cross-reactivity The kit includes a 96-well plate with capture antibody detection antibody read buffer wash buffer calibrator and positive control
Catalog No. 50-000-81379 Supplier Meso Scale Discovery Supplier No. K1526HK21
The Meso Scale Discovery U-PLEX Mouse Insulin Assay is an electrochemiluminescent immunoassay for measuring insulin in mouse serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive accurate results in 3 5 hours The assay detects as low as 50 pg/mL and includes a calibrator for a standard curve from 2-10 000 pg/mL It requires 25 L of sample and offers high specificity without cross-reactivity The kit includes a 96-well plate with capture antibody detection antibody read buffer wash buffer calibrator and positive control