Learn More
Meso Scale Discovery U-PLEX Mouse Insulin ASYQP
SDP
Catalog No. 5000082175
Encompass
The Meso Scale Discovery U-PLEX Mouse Insulin Assay is an electrochemiluminescent immunoassay for the quantitative measurement of insulin in mouse serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive accurate and reproducible results in just 3 5 hours The assay has a lower limit of detection of 50 pg/mL and includes a 96-well plate pre-coated with capture antibodies detection antibodies conjugated to MSD SULFO-TAG calibrators and controls for reliable quantitation This assay allows high-throughput analysis of insulin levels in mouse samples
Catalog No. 50-000-82175 Supplier Meso Scale Discovery Supplier No. K1526HK22
Add to Cart
Add to Cart
The Meso Scale Discovery U-PLEX Mouse Insulin Assay is an electrochemiluminescent immunoassay for the quantitative measurement of insulin in mouse serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive accurate and reproducible results in just 3 5 hours The assay has a lower limit of detection of 50 pg/mL and includes a 96-well plate pre-coated with capture antibodies detection antibodies conjugated to MSD SULFO-TAG calibrators and controls for reliable quantitation This assay allows high-throughput analysis of insulin levels in mouse samples